Orders LinkContactUsLinkComponent
ISE standard solutions
Supplier:   WTW
ISE standard solutions
Supplier:  WTW
Certificates
ISE standard solutions
ISE standard solutions
6 Options Available Below
Product Details & Documents
These standard solutions for calibration contain 10 g of the ion to be measured per 10 liters.
Product Family Options
Product Information
  • Descrizione
    Conf
    Availability
    Price
  • Soluzione tampone ES/Cl: 10 g/l Cl (Cl800)
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 234.00
    SFr. 234.00
    1 * 1 L
       SFr. 234.00
  • Soluzione tampone ES/F: 10 g/l F (F800)
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 234.00
    SFr. 234.00
    1 * 1 L
       SFr. 234.00
  • Standard solution calcium ions (ES/Ca)
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 234.00
    SFr. 234.00
    1 * 1 L
       SFr. 234.00
  • Standard solution potassium ions (ES/K)
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 234.00
    SFr. 234.00
    1 * 1 L
       SFr. 234.00
  • Standard solution nitrate (ES/NO₃)
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 234.00
    SFr. 234.00
    1 * 1 L
       SFr. 234.00
  • Standard solution sodium ions (ES/Na)
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 234.00
    SFr. 234.00
    1 * 1 L
       SFr. 234.00