Track sets
Track set, length 3962,4 mm
Codice catalogo METOTTS13NA
Nome del fornitore: Metro International
Vai a
Product Details & Documents
Track sets includes necessary sections of track for assembling track runs up to 6400 mm. Only one track set is required between stationary units.
Specifiche
- Descrizione:Track set, length 3962,4 mm
- Seq:05