Electrode stand with swivel arm, Eutech
Catalog # EUTEECPHELSTDC
Supplier: EUTECH
Specifications
- Per:Up to four standard size electrodes and one ATC probe
- Descrizione:Electrode stand with swivel arm
- Environmentally Preferable:
- Conf:1
Specifications
About this item
This electrode stand with swivel arm allows convenient hands-free operation. Holds up to four standard size electrodes and one ATC probe.
- Weighted 6-inch diameter base allows users to pivot holder 360° for more flexibility
- Arm allows users to move electrode side to side or up and down while keeping electrode at a constant vertical angle