Viscosity test standard for PTP, VHG
Catalog # PROOG-VPTPVISC-50
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Conf:1
- Descrizione:Viscosity test standard, 40 °C and 100 °C
Specifications
About this item
This proficiency testing program (PTP) was specially designed to give analytical laboratories instant validation while measuring viscosity in new and used oils.
Certifications: Traceable to CRM. ISO/IEC 17043 accredited.