Volumetric flasks, class A, glass, clear, ISO-thread and screw caps
Supplier: NEUBERT VOLUME GLASSWAERE
Certificates
About this item
Borosilicate glass with ISO thread and screw cap
- Conformity certified with dated batch identification
- Screw cap made of Celanex red with PTFE gasket
- With one graduation mark
Frequently Bought Together