Aspirator bottles
Catalog # BEHRQL9002607
Supplier: BEHR
Specifications
- Ø est:210 mm
- Capacità:10000 ml
- Altezza:427 mm
- Conf:1
Specifications
About this item
Aspirator bottles, HDPE natural, narrow-neck storage bottles with scale.
- UV protection for long life
- Resistant to acids and chemicals
- Special colour and printing possible
Storage vessel with drain made of food-safe polyethylene.