Orders LinkContactUsLinkComponent
Cyclosporine, Reference Standards
Cyclosporine, Reference Standards
Nome del fornitore:   USP
Cyclosporine, Reference Standards
Nome del fornitore:  USP
Certificates
2 varianti
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Cyclosporine Resolution Mixture: This material is an approximate 100:1 (w/w) mixture of cyclosporine and cyclosporine U
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifiche
Specifiche
  • Catalog No:
  • 1158504.
  • 1158650.
  • Descrizione:
  • Cyclosporine
  • Cyclosporine Resolution Mixture
  • CAS No.:
  • 59865-13-3
  • Condizioni di conservazione:
  • Refrigerator
Specifiche
Product Family Options
Informazioni sul prodotto
  • Standard
    Tipo di conf.
    Conf
    Disponibilità
    Price
  • 1158504.
    Cyclosporine
    Cyclosporine
    USP reference standard
    Bottiglia di vetro
    50 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 50 mg
    1 * 50 mg - 1 350.00 SFr.
    1 350.00 SFr.
    1 * 50 mg
       1 350.00 SFr.
    Specifiche:
    More
    Specifications
    Cat. No.
    1158504.
    Descrizione
    Cyclosporine
    CAS No.
    59865-13-3
    Condizioni di conservazione
    Refrigerator
  • 1158650.
    Cyclosporine Resolution Mixture
    Cyclosporine Resolution Mixture
    USP reference standard
    Vial
    25 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 25 mg
    1 * 25 mg - 1 430.00 SFr.
    1 430.00 SFr.
    1 * 25 mg
       1 430.00 SFr.
    Specifiche:
    More
    Specifications
    Cat. No.
    1158650.
    Descrizione
    Cyclosporine Resolution Mixture
    CAS No.
    Condizioni di conservazione