Orders LinkContactUsLinkComponent
Levocetirizine, Reference Standards
 :   USP
Levocetirizine, Reference Standards
 :  USP
Levocetirizine, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
 : US Pharmacopea Reference Standard
  • Catalog No:
  • 1359969.
  • 1359958.
  • Descrizione:
  • Levocetirizine Dihydrochloride
  • Levocetirizine Amide
  • CAS No.:
  • 130018-87-0
  • 909779-33-5
  • Formula:
  • C₂₁H₂₇Cl₃N₂O₃
  • C₂₄H₃₆N₄O₅
Product Family Options
  • Descrizione
    Standard
    Tipo di conf.
    Conf
  • Levocetirizine Dihydrochloride
    USP reference standard
    Vial
    200 mg
    1 * 200 mg
    1 * 200 mg - SFr. 425.00
    SFr. 425.00
    1 * 200 mg
       SFr. 425.00
    Specifications
    Cat. No.
    1359969.
    Descrizione
    Levocetirizine Dihydrochloride
    CAS No.
    130018-87-0
    Formula
    C₂₁H₂₇Cl₃N₂O₃
  • Levocetirizine Amide
    USP reference standard
    Vial
    75 mg
    1 * 75 mg
    1 * 75 mg - SFr. 1’350.00
    SFr. 1’350.00
    1 * 75 mg
       SFr. 1’350.00
    Specifications
    Cat. No.
    1359958.
    Descrizione
    Levocetirizine Amide
    CAS No.
    909779-33-5
    Formula
    C₂₄H₃₆N₄O₅