Orders LinkContactUsLinkComponent
Liothyronine, Reference Standards
Liothyronine, Reference Standards
Nome del fornitore:   USP
Liothyronine, Reference Standards
Nome del fornitore:  USP
Certificates
2 varianti
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Documents
Specifications for Product Family
Specifiche
Specifiche
  • Catalog No:
  • 1368008.
  • 1368019.
  • Descrizione:
  • Liothyronine
  • Liothyronine Sodium
  • CAS No.:
  • 6893-02-3
  • 55-06-1
  • Formula:
  • C₁₅H₁₂I₃NO₄
Specifiche
Product Family Options
Informazioni sul prodotto
  • Standard
    Tipo di conf.
    Conf
    Disponibilità
    Price
  • Liothyronine
    USP reference standard
    Vial
    250 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 250 mg
    1 * 250 mg - SFr. 462.00
    SFr. 462.00
    1 * 250 mg
       SFr. 462.00
    Specifiche:
    More
    Specifications
    Cat. No.
    1368008.
    Descrizione
    Liothyronine
    CAS No.
    6893-02-3
    Formula
    C₁₅H₁₂I₃NO₄
  • 1368019.
    Liothyronine Sodium
    Liothyronine Sodium
    USP reference standard
    Vial
    200 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 200 mg
    1 * 200 mg - SFr. 770.00
    SFr. 770.00
    1 * 200 mg
       SFr. 770.00
    Specifiche:
    More
    Specifications
    Cat. No.
    1368019.
    Descrizione
    Liothyronine Sodium
    CAS No.
    55-06-1
    Formula
Consigli