Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Mouse recombinant OX40

Mouse recombinant OX40

50 µG
Codice catalogo PRSI92-548
Nome del fornitore:   ProSci Inc.
Mouse recombinant OX40
Codice catalogo PRSI92-548
Nome del fornitore:  ProSci Inc.
Numero CAS:  
Mouse recombinant OX40

50 µG

Numero CAS:  
Price
SFr. 430.00
Log in per disponibilità
1 * 50 µG
Product Details & Documents

OX40, also termed CD134 and TNFRSF4, is a T cell co-stimulatory molecule of the TNF receptor superfamily which plays a key role in the survival and homeostasis of effector and memory T cells. OX40 is expressed on CD4+ and CD8+ T cells upon engagement of the TCR by antigen presenting cells along with co-stimulation by CD40-CD40 Ligand and CD28-B7. The interaction between OX40 and OX40 ligand (OX40L) will occur when activated T cells bind to professional antigen-presenting cells (APCs). The T-cell functions, including cytokine production, expansion, and survival, are then enhanced by the OX40 costimulatory signals. OX40 signals are critical for controllling the function and differentiation of Foxp3+ regulatory T cells. OX40-OX40L interaction regulates T-cell tolerance, peripheral T-cell homeostasis, and T-cell-mediated inflammatory diseases.

Fusion-Tag: N-Fc tag

This recombinant protein can be used for biological assays. For research use only.

Documents

Specifiche

Specifiche

  • Conf:
    50 µG
  • Peso molecolare:
    47.4 kD
  • Cat. No.:
    PRSI92-548
  • Condizioni di conservazione:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    OX40
  • Purezza:
    > 95% as determined by reducing SDS-PAGE
  • Protein synonyms:
    REF: C1125|OX40|Tumor necrosis factor receptor superfamily member 4|Txgp1|Tnfrsf4|CD134
  • Specie:
    Mouse
  • Sequenza:
    Val20-Pro211
  • Formulazione:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Sequenza tag:
    N-Fc tag

Specifiche

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..