Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant FGF17 (from E. coli)
Human recombinant FGF17 (from E. coli)
Codice catalogo PRSI92-171
Nome del fornitore:  ProSci Inc.
Numero CAS:  
Human recombinant FGF17 (from E. coli)
Codice catalogo PRSI92-171
Nome del fornitore:  ProSci Inc.
Numero CAS:  

Specifiche

  • Conf:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Sorgente luminosa:
    E. coli
  • Specie:
    Human
  • Condizioni di conservazione:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    FGF17|Fibroblast Growth Factor 17|FGF-17
  • Protein/peptide name:
    FGF17
  • Purezza:
    > 95% as determined by reducing SDS-PAGE
  • Peso molecolare:
    22.6 kD
  • Sequenza:
    Thr23-Thr216
  • Formulazione:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Applicazioni testate:
    Bioassay

Specifiche

Informazioni su questo articolo

Fibroblast Growth Factor 17 (FGF17) is a member of the heparin-binding growth factors family that is prominently expressed in the cerebellum and cortex. Proteins of this family possess broad mitogenic and cell survival activities and they are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumour growth, and invasion. FGF17 plays an important role in the regulation of embryonic development and it acts as signalling molecule in the induction and patterning of the embryonic brain. In addition, FGF17 stimulates the proliferation and activation of cells that express FGF receptors.

  • This recombinant protein can be used for biological assays. For research use only.

This recombinant protein can be used for biological assays. For research use only.