Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant CCL21A (from E. coli)
Mouse recombinant CCL21A (from E. coli)
Catalog # PRSI92-158
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant CCL21A (from E. coli)
Catalog # PRSI92-158
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Conf:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Sorgente luminosa:
    E. coli
  • Specie:
    Mouse
  • Condizioni di conservazione:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Scya21|TCA4|Beta-Chemokine Exodus-2|6Ckine|Scya21a|Thymus-Derived Chemotactic Agent 4|C-C Motif Chemokine 21a|Small-Inducible Cytokine A21a|Ccl21a
  • Protein/peptide name:
    CCL21A
  • Purezza:
    > 95% as determined by reducing SDS-PAGE
  • Peso molecolare:
    12 kD
  • Sequenza:
    Ser24-Gly133
  • Formulazione:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Applicazioni testate:
    Bioassay

Specifications

About this item

C-C Motif Chemokine 21a (CCL21a) is a small secreted cytokine that belongs to the intercrine beta (CC chemokine) family. Mouse CCL21 cDNA encodes a 133 amino acid residue protein with a 23 residue signal peptide that is cleaved to generate the 110 residue mature protein. Mouse CCL21 has three forms while CCL21a has Ser-65. CCL21 elicits its effects by binding to a cell surface chemokine receptor known as CCR7 and CXCR3. Mouse CCL21 inhibits hemopoiesis and stimulates chemotaxis. It has chemotactic function in vitro for thymocytes and activated T-cells, but not for B-cells, macrophages, or neutrophils. Mouse CCL21 shows preferential activity towards naive T-cells and may play a role in mediating homing of lymphocytes to secondary lymphoid organs.

This recombinant protein can be used for biological assays. For research use only.