Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Human recombinant GADD45B (from E. coli)

50 µG
Codice catalogo PRSI92-003
Nome del fornitore:   ProSci Inc.
Human recombinant GADD45B (from <i>E. coli</i>)
Codice catalogo PRSI92-003
Nome del fornitore:  ProSci Inc.
Numero CAS:  
Human recombinant GADD45B (from E. coli)

50 µG

Numero CAS:  
Price
752.00 SFr.
Log in per disponibilità
1 * 50 µG
Product Details & Documents

Growth Arrest and DNA Damage-Inducible Protein GADD45 beta (GADD45B) is a member of the GADD45 family. GADD45B has been shown to interact with MAP3K4, ASK1, MAP2K7, and GADD45GIP1. GADD45B is involved in the regulation of growth and apoptosis. GADD45B reacts to environmental stresses by mediating activation of stress-responsive MTK1/MEKK4 kinase, which is an upstream activator of both p38 and JNK MAPKs. In addition, GADD45B participates in the down-regulation of hepatocellular carcinoma (HCC). It may serve as a possible therapeutic target.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

Specifiche

Specifiche

  • Conf:
    50 µG
  • Protein/peptide name:
    GADD45B
  • Purezza:
    > 95% as determined by reducing SDS-PAGE
  • Protein synonyms:
    GADD45B|Negative Growth Regulatory Protein MyD118|Myeloid Differentiation Primary Response Protein MyD118|MYD118|Growth Arrest and DNA Damage-Inducible Protein GADD45 Beta
  • Specie:
    Human
  • Sequenza:
    Met1-Arg160
  • Sorgente luminosa:
    E. coli
  • Peso molecolare:
    20 kD
  • Cat. No.:
    PRSI92-003
  • Condizioni di conservazione:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein/peptide type:
    Recombinant
  • Applicazioni testate:
    Bioassay
  • Formulazione:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Sequenza tag:
    N-6 His tag

Specifiche

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.