ContactUsLinkComponent
Human recombinant ST8SIA1 (from cells)
Catalog # PRSI91-569
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant ST8SIA1 (from cells)
Catalog # PRSI91-569
Supplier:  ProSci Inc.
CAS Number:  
Specifications
  • Conf:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Sorgente luminosa:
    Cells
  • Specie:
    Human
  • Sequenza tag:
    C-6 His tag
  • Condizioni di conservazione:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Ganglioside GT3 Synthase|8-Sialyltransferase|Sialyltransferase 8A|Sialytransferase St8Sia I|SIAT8|8-Sialyltransferase 8A|ST8SIA1|SIAT8-A|ST8SiaI|Alpha-2|SIAT8A|Ganglioside GD3 Synthase|Alpha-N-Acetylneuraminide Alpha-2
  • Protein/peptide name:
    ST8SIA1
  • Purezza:
    > 95% as determined by reducing SDS-PAGE
  • Peso molecolare:
    36.2 kD
  • Sequenza:
    Tyr49-Ser356
  • Formulazione:
    Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl,150mM NaCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Applicazioni testate:
    Bioassay
Specifications
About this item
alpha-N-Acetylneuraminide alpha-2,8-Sialyltransferase (ST8SIA1) belongs to the glycosyltransferase 29 family. ST8SIA1 is a sialytransferase that catalyzes the transfer of sialic acid from CMP-sialic acid to GM3 to produce GD3 and GT3. ST8SIA1 is highly expressed in melanoma cell lines, adult and fetal brain, low expressed in adult and fetal lung. ST8SIA1 may act as a type II transmembrane protein with a short N-termianl cytoplasmic domain and a single-pass transmembrane domain folowed by an enzymatic domain in the lument of the Golgi apparatus.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag