Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant C1qTNF1 (from Cells)
Human recombinant C1qTNF1 (from Cells)
Catalog # PRSI91-284
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant C1qTNF1 (from Cells)
Catalog # PRSI91-284
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Conf:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Sorgente luminosa:
    Cells
  • Specie:
    Human
  • Sequenza tag:
    C-6 His tag
  • Condizioni di conservazione:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    G Protein-Coupled Receptor-Interacting Protein|CTRP1|Complement C1q Tumor Necrosis Factor-Related Protein 1|GIP|C1QTNF1
  • Protein/peptide name:
    C1qTNF1
  • Purezza:
    > 95% as determined by reducing SDS-PAGE
  • Peso molecolare:
    30.24 kD
  • Sequenza:
    Arg26-Pro281
  • Formulazione:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Applicazioni testate:
    Bioassay

Specifications

About this item

C1QTNF1 is a secreted protein,contains 1 C1q domain and 1 collagen-like domain. C1qTNF proteins constitute a highly conserved family of Acrp30/Adiponectin paralogs that share a modular organization comprising an N-terminal signal peptide, a short variable region, a collagenous domain and a C-terminal globular domain. C1qTNF proteins are predicted to have trimeric structures that assemble into hexameric and higher order molecular forms. C1QTNF1 is a novel adipokine, providing a significant framework to further address the physiological functions and mechanisms of the action of this family of secreted glycoproteins in normal and disease states. C1QTNF1 increases the production of aldosterone. C1QTNF1 is vastly expressed in obese subjects as well as up-regulated in hypertensive patients, C1QTNF1 is identified molecular link between obesity and hypertension. C1QTNF1 expression may be associated with a low-grade chronic inflammation status in adipose tissues.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag