ContactUsLinkComponent
Human Recombinant EIF1AX (from E. coli)
Human Recombinant EIF1AX (from E. coli)
Catalog # PRSI91-199
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant EIF1AX (from E. coli)
Catalog # PRSI91-199
Supplier:  ProSci Inc.
CAS Number:  
Specifications
  • Conf:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Sorgente luminosa:
    E. coli
  • Specie:
    Human
  • Sequenza tag:
    N-6 His tag
  • Condizioni di conservazione:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Eukaryotic Translation Initiation Factor 1A X-Chromosomal|EIF1AX|eIF-1A X Isoform|EIF1A|Eukaryotic Translation Initiation Factor 4C|eIF-4C|EIF4C
  • Protein/peptide name:
    EIF1AX
  • Purezza:
    > 95% as determined by reducing SDS-PAGE
  • Peso molecolare:
    18.6 kD
  • Sequenza:
    Met1-Ile144
  • Formulazione:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Applicazioni testate:
    Bioassay
Specifications
About this item
Eukaryotic Translation Initiation Factor 1A, X-Chromosomal (EIF1AX) is an essential eukaryotic translation initiation factor that belongs to the eIF-1A family. EIF1AX is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA and has been shown to interact with IPO13. EIF1AX contains one S1-like domain and seems to be required for maximal rate of protein biosynthesis. Enhances ribosome dissociation into subunits and stabilizes the binding of the initiator Met-tRNA(I) to 40 S ribosomal subunits.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag