Human IKK epsilon peptide (16 aa near C-terminus)
Catalog # PRSI2329P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Conf:50 µG
- Specie:Human
- Condizioni di conservazione:Store at –20 °C, stable for one year.
- Numero amminoacido:16 aa near C-terminus
- Protein/peptide name:IKK epsilon Peptide (16 aa near C-terminus)
- Applicazioni testate:Blocking
Specifications
About this item
IKK epsilon peptide is used for blocking the activity of IKK epsilon antibody.