Specifications
- Tipo di anticorpi:Nucleobindin 1
- Simbolo dell'antigene:NUCB1
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Rabbit
- Reattività:Human
- Forma:Lyophilized
- Immunogeno:A synthetic KLH-conjugated peptide corresponding a region of human NUCB1.
- Taglia:100 µg
- Conf:100 µG
Specifications
About this item
Calnuc or NUCB1 belongs to the nucleobindin family. It is a major calcium-binding protein of the Golgi and is a good Golgi marker. It may be involved in calcium homeostasis. Calnuc also plays roles in regulation of levels of.-amyloid precursor protein (APP) and its proteolytic metabolites to further affect the patho/physiological functions of APP including Alzheimer's disease pathogenesis. Useful for immunohistochemistry and western blot.
Antibody is purified by Protein A/G affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NUCB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human