Anti-SIGLEC9 Rabbit Polyclonal Antibody
PRSI30-257
- Tipo di anticorpi:Sialic Acid Binding Ig-like Lectin 9
- Simbolo dell'antigene:SIGLEC9
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Rabbit
- Reattività:Human
- Western blot:Yes
- Forma:Lyophilized
- Sinomini antigene:CDw329|FOAP-9|siglec-9|OBBP-LIKE|CD329
- Immunogeno:A synthetic peptide corresponding a region of human SIGLEC9.
- Taglia:50 µg
- Conf:50 µG
SIGLEC9 is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It preferentially binds to alpha2,3- or 2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SIGLEC9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human