- Tipo di anticorpi:Ts translation elongation Factor, mitochondrial
- Simbolo dell'antigene:TSFM
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Rabbit
- Reattività:Human
- Western blot:Yes
- Forma:Lyophilized
- Sinomini antigene:EFTSMT|EFTS
- Immunogeno:A synthetic peptide corresponding a region of human TSFM.
- Taglia:50 µg
- Conf:50 µG
The TSFM protein is a mitochondrial translation elongation factor. Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes.Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes. The TSFM gene encodes a mitochondrial translation elongation factor (Smeitink et al., 2006 [PubMed 17033963]).
TSFM antibody can be used for detection of TSFM by ELISA at 1:312500. TSFM antibody can be used for detection of TSFM by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TSFM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human