ContactUsLinkComponent
Drug and metabolite standards, dexamethasone, LoGiCal®
Drug and metabolite standards, dexamethasone, LoGiCal®
Nome del fornitore:  LGC Standards PROMOCHEM
Certificati
Drug and metabolite standards, dexamethasone, LoGiCal®
Nome del fornitore:  LGC Standards PROMOCHEM
Specifiche
  • CAS No.:
    1177-87-3
  • Formula:
    C₂₄H₃₁FO₆
  • Descrizione:
    Dexamethasone acetate