Carbidopa, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
- Catalog No:
- 1095506.
- 1095517.
- Descrizione:
- Carbidopa
- Carbidopa Related Compound A
- Synonym(s):
- 3-O-Methylcarbidopa
- CAS No.:
- 38821-49-7
- 85933-19-3
- Formula:
- C₁₀H₁₄N₂O₄
- C₁₁H₁₆N₂O₄
Product Family Options
- DescrizioneStandardTipo di conf.Conf
- CarbidopaUSP reference standardVial400 mg1 * 400 mgSFr. 462.00
Specifications
Cat. No.1095506.DescrizioneCarbidopaSynonym(s)CAS No.38821-49-7FormulaC₁₀H₁₄N₂O₄ - Carbidopa Related Compound AUSP reference standardVial25 mg1 * 25 mgSFr. 1’480.00
Specifications
Cat. No.1095517.DescrizioneCarbidopa Related Compound ASynonym(s)3-O-MethylcarbidopaCAS No.85933-19-3FormulaC₁₁H₁₆N₂O₄