Orders LinkContactUsLinkComponent
Capecitabine, Reference Standards
Nome del fornitore:   USP
Capecitabine, Reference Standards
Nome del fornitore:  USP
Certificates
4 varianti
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
 : US Pharmacopea Reference Standard
Specifications for Product Family
Specifiche
Specifiche
  • Catalog No:
  • 1090706.
  • 1090717.
  • 1090728.
  • 1090739.
  • Descrizione:
  • Capecitabine
  • Capecitabine Related Compound A
  • Capecitabine Related Compound B
  • Capecitabine Related Compound C
  • Synonym(s):
  • 5'-deoxy-5-fluorocytidine
  • 5'-deoxy-5-fluorouridine
  • 2',3'-O-carbonyl-5'-deoxy-5-fluoro-N4-(pentyloxycarbonyl)cytidine
  • CAS No.:
  • 154361-50-9
  • 66335-38-4
  • 3094-09-5
  • 921769-65-5
  • Formula:
  • C₁₅H₂₂FN₃O₆
  • C₉H₁₂FN₃O₄
  • C₉H₁₁FN₂O₅
  • C₁₆H₂₀FN₃O₇
Specifiche
Product Family Options
Informazioni sul prodotto
  • Standard
    Tipo di conf.
    Conf
    Disponibilità
    Price
  • 1090706.
    Capecitabine
    Capecitabine
    USP reference standard
    Vial
    200 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 200 mg
    1 * 200 mg - 450.00 SFr.
    450.00 SFr.
    1 * 200 mg
       450.00 SFr.
    Specifiche:
    More
    Specifications
    Cat. No.
    1090706.
    Descrizione
    Capecitabine
    Synonym(s)
    CAS No.
    154361-50-9
    Formula
    C₁₅H₂₂FN₃O₆
  • 1090717.
    Capecitabine Related Compound A
    Capecitabine Related Compound A
    USP reference standard
    Vial
    15 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 15 mg
    1 * 15 mg - 1 350.00 SFr.
    1 350.00 SFr.
    1 * 15 mg
       1 350.00 SFr.
    Specifiche:
    More
    Specifications
    Cat. No.
    1090717.
    Descrizione
    Capecitabine Related Compound A
    Synonym(s)
    5'-deoxy-5-fluorocytidine
    CAS No.
    66335-38-4
    Formula
    C₉H₁₂FN₃O₄
  • 1090728.
    Capecitabine Related Compound B
    Capecitabine Related Compound B
    USP reference standard
    Vial
    20 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 20 mg
    1 * 20 mg - 1 450.00 SFr.
    1 450.00 SFr.
    1 * 20 mg
       1 450.00 SFr.
    Specifiche:
    More
    Specifications
    Cat. No.
    1090728.
    Descrizione
    Capecitabine Related Compound B
    Synonym(s)
    5'-deoxy-5-fluorouridine
    CAS No.
    3094-09-5
    Formula
    C₉H₁₁FN₂O₅
  • 1090739.
    Capecitabine Related Compound C
    Capecitabine Related Compound C
    USP reference standard
    Vial
    10 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 10 mg
    1 * 10 mg - 1 440.00 SFr.
    1 440.00 SFr.
    1 * 10 mg
       1 440.00 SFr.
    Specifiche:
    More
    Specifications
    Cat. No.
    1090739.
    Descrizione
    Capecitabine Related Compound C
    Synonym(s)
    2',3'-O-carbonyl-5'-deoxy-5-fluoro-N4-(pentyloxycarbonyl)cytidine
    CAS No.
    921769-65-5
    Formula
    C₁₆H₂₀FN₃O₇