Containment berms, QuickBerm® Plus
Informazioni su questo articolo
The QuickBerm Plus® offers superior protection and chemical resistance against the harshest of liquids, including petroleum, oils, greases, and most acids.
- Single-piece construction - patented Rigid-Lock wall-support system integral to berm provides unobstructed work space
- Quick and easy setup, no assembly required
- Withstands long-term UV exposure and extreme temperatures
- Finished hem edge on top of sidewall prevents fraying and adds strength
A simple push on the walls unlocks the support and lays the wall down for drive-through capability from any angle. Reinforced CriticalCorner™ design with wraparound diagonal radio frequency (RF) welds offer exceptionally strong structural integrity to prevent seam leaks, provide fluid-tight reliability, and extend berm life expectancy.
For added resistance against rocky surfaces or heavy in-out traffic use tarps, ground mats, track mats, or track runners. Excellent for heavy vehicle traffic
Certifications: Helps to comply with EPA for containment and spill prevention (SPCC)
Informazioni per l'ordine: Anchors not included.
6 varianti
Product Details & Documents
The QuickBerm Plus® offers superior protection and chemical resistance against the harshest of liquids, including petroleum, oils, greases, and most acids.
- Single-piece construction - patented Rigid-Lock wall-support system integral to berm provides unobstructed work space
- Quick and easy setup, no assembly required
- Withstands long-term UV exposure and extreme temperatures
- Finished hem edge on top of sidewall prevents fraying and adds strength
A simple push on the walls unlocks the support and lays the wall down for drive-through capability from any angle. Reinforced CriticalCorner™ design with wraparound diagonal radio frequency (RF) welds offer exceptionally strong structural integrity to prevent seam leaks, provide fluid-tight reliability, and extend berm life expectancy.
For added resistance against rocky surfaces or heavy in-out traffic use tarps, ground mats, track mats, or track runners. Excellent for heavy vehicle traffic
Certifications: Helps to comply with EPA for containment and spill prevention (SPCC)
Informazioni per l'ordine: Anchors not included.
Product Family Options
Informazioni sul prodotto
- CapacitàL×P×ADisponibilitàPrice
- 119-0358Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 24 kgRigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 24 kg2252 l3000×2603×305 mmLog in per disponibilità1 * 1 ST2 580.00 SFr.
- 119-0359Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 37,6 kgRigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 37,6 kg4240 l4600×3000×305 mmLog in per disponibilità1 * 1 ST3 690.00 SFr.
- 119-0360Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 68,0 kgRigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 68,0 kg9501 l8500×3700×305 mmLog in per disponibilità1 * 1 ST6 330.00 SFr.
- 119-0361Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 131,5 kgRigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 131,5 kg16978 l15200×3700×305 mmLog in per disponibilità1 * 1 ST8 690.00 SFr.
- 119-0362Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 139,3 kgRigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 139,3 kg18321 l16500×3700×305 mmLog in per disponibilità1 * 1 ST9 120.00 SFr.
- 119-0363Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 141,1 kgRigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 141,1 kg22183 l17100×4300×305 mmLog in per disponibilità1 * 1 ST10 140.00 SFr.