Orders LinkContactUsLinkComponent
Raloxifene Related Compounds, Reference Standard
Raloxifene Related Compounds, Reference Standard
Nome del fornitore:   USP
Raloxifene Related Compounds, Reference Standard
Nome del fornitore:  USP
Certificates
2 varianti
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifiche
Specifiche
  • Catalog No:
  • 1598201.
  • 1598234.
  • Descrizione:
  • Raloxifene Hydrochloride
  • Raloxifene Related Compound C
  • Synonym(s):
  • 1-(2-{4-[6-hydroxy-2-(4-hydroxyphenyl)benzothiophene-3-carbonyl]phenoxy}ethyl)piperidine 1-oxide monohydrate
  • CAS No.:
  • 82640-04-8
  • 195454-31-0
  • Formula:
  • C₂₈H₂₈ClNO₄S
  • C₂₈H₂₇NO₅S
  • N. ONU:
  • 3077
Specifiche
Product Family Options
Informazioni sul prodotto
  • Standard
    Tipo di conf.
    Conf
    Disponibilità
    Price
  • 1598201.
    Raloxifene Hydrochloride
    Raloxifene Hydrochloride
    USP reference standard
    Vial
    200 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 200 mg
    1 * 200 mg - SFr. 420.00
    SFr. 420.00
    1 * 200 mg
       SFr. 420.00
    Specifiche:
    More
    Specifications
    Cat. No.
    1598201.
    Descrizione
    Raloxifene Hydrochloride
    Synonym(s)
    CAS No.
    82640-04-8
    Formula
    C₂₈H₂₈ClNO₄S
    N. ONU
    3077
  • 1598234.
    Raloxifene Related Compound C
    Raloxifene Related Compound C
    USP reference standard
    Vial
    20 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 20 mg
    1 * 20 mg - SFr. 1’370.00
    SFr. 1’370.00
    1 * 20 mg
       SFr. 1’370.00
    Specifiche:
    More
    Specifications
    Cat. No.
    1598234.
    Descrizione
    Raloxifene Related Compound C
    Synonym(s)
    1-(2-{4-[6-hydroxy-2-(4-hydroxyphenyl)benzothiophene-3-carbonyl]phenoxy}ethyl)piperidine 1-oxide monohydrate
    CAS No.
    195454-31-0
    Formula
    C₂₈H₂₇NO₅S
    N. ONU