Syringes, Cavro
Cavro XL syringe with Luer lock termination, 1 ml
Codice catalogo SGEA008140
Nome del fornitore: Trajan Scientific and Medical
Vai a
Product Details & Documents
These cavro syringes are a type of instrument syringes used in chromatography.
- Luer lock termination
- Gas tight plunger with CAVRO XL mounting hole
Specifiche
- Volume:1 ml
- Descrizione:Cavro XL syringe with Luer lock termination
- Seq:01