Drums, stackable, with tamper evident cap
Drums, with tamper evident cap, 25,550 ml
Codice catalogo DELT206025
Nome del fornitore: DELTALAB
Vai a
Product Details & Documents
Stackable drums, made of HDPE, natural.
- Hermetical closure
- For alimentary use
They embody a handle and a tamper evident, ribbed screw red cap, made of HDPE, with internal joint and internal security ring.
Specifiche
- Capacità:25,550 ml
- Descrizione:Drums, with tamper evident cap