Orders LinkContactUsLinkComponent
Ofloxacin, Reference Standards
Ofloxacin, Reference Standards
Nome del fornitore:   USP
Ofloxacin, Reference Standards
Nome del fornitore:  USP
Certificates
3 varianti
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifiche
Specifiche
  • Catalog No:
  • 1478119.
  • 1478108.
  • 1478083.
  • Descrizione:
  • Ofloxacin Related Compound A
  • Ofloxacin
  • R-Ofloxacin
  • Synonym(s):
  • 9-Fluoro-3-methyl-7-oxo-10-(piperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid
  • (R)-9-fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de][1,4]benzoxazine-6-carboxylic acid
  • CAS No.:
  • 82419-52-1
  • 82419-36-1
  • 100986-86-5
  • Formula:
  • C₁₇H₁₈FN₃O₄
  • C₁₈H₂₀FN₃O₄
  • C₁₈H₂₀FN₃O₄
Specifiche
Product Family Options
Informazioni sul prodotto
  • Standard
    Tipo di conf.
    Conf
    Disponibilità
    Price
  • 1478119.
    Ofloxacin Related Compound A
    Ofloxacin Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 25 mg
    1 * 25 mg - SFr. 1’440.00
    SFr. 1’440.00
    1 * 25 mg
       SFr. 1’440.00
    Specifiche:
    More
    Specifications
    Cat. No.
    1478119.
    Descrizione
    Ofloxacin Related Compound A
    Synonym(s)
    9-Fluoro-3-methyl-7-oxo-10-(piperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid
    CAS No.
    82419-52-1
    Formula
    C₁₇H₁₈FN₃O₄
  • 1478108.
    Ofloxacin
    Ofloxacin
    USP reference standard
    Vial
    200 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 200 mg
    1 * 200 mg - SFr. 462.00
    SFr. 462.00
    1 * 200 mg
       SFr. 462.00
    Specifiche:
    More
    Specifications
    Cat. No.
    1478108.
    Descrizione
    Ofloxacin
    Synonym(s)
    CAS No.
    82419-36-1
    Formula
    C₁₈H₂₀FN₃O₄
  • 1478083.
    R-Ofloxacin
    R-Ofloxacin
    USP reference standard
    Vial
    25 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 25 mg
    1 * 25 mg - SFr. 1’310.00
    SFr. 1’310.00
    1 * 25 mg
       SFr. 1’310.00
    Specifiche:
    More
    Specifications
    Cat. No.
    1478083.
    Descrizione
    R-Ofloxacin
    Synonym(s)
    (R)-9-fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de][1,4]benzoxazine-6-carboxylic acid
    CAS No.
    100986-86-5
    Formula
    C₁₈H₂₀FN₃O₄