Certified reference material, soya-maize oil blend, BCR®
PROOBCR-162R
:
- Tipo di conf.:Ampoule, amber
- Conf:5,5 g
- Descrizione:Soya-maize oil blend - fatty acids
A unit contains approximately 5,5 gm soya-maize oil blend in 10 ml amber ampoule.
- Fatty acid analysis
: IRMM certified.
: Store in Freezer below –20 °C.