Organic reference standards, ivermectin standard, BP
Catalog # PROOBP864
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Conf:250 mg
- Standard:Reference standard
- CAS No.:70288-86-7
- Descrizione:Ivermectin
- Formula:C₄₈H₇₄O₁₄
- Descrizione:Ivermectin