VWR®, Silane Coated, Double Frosted Slides
Catalog # 631-1162
Supplier: VWR Collection
Specifications
- L×P:76×25 mm
- Descrizione:Double-frosted silane coated slides
- Seq:007
- Spessore:1,0 (-0,05/+0,1) mm
- Conf:2.500
Specifications
About this item
Silane (aminoalkylsilane) coating provide permanent adhesion, which can bind tissue sections to glass slides with a sticky surface.
- Smooth edges for operational safety
- Marking with pencils, marker pens and conventional labelling systems
- Shelf life of 18 months
Recommended for H&E stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopathology examination.
Packaging: 50 pieces per box. 50 boxes per case.
Caution: Fragile items should be stored in dry and proper laboratory temperature conditions.