Organic reference standards, Mebendazole, BP
Catalog # PROOBP1008
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Conf:10 mg
- Standard:Reference Standard
- CAS No.:31431-39-7
- Descrizione:Mebendazole, BP 1008
- Environmentally Preferable:
- Formula:C₁₆H₁₃N₃O₃
- Descrizione:Mebendazole