Cut resistant gloves, ChainExtra, with long cuff
Supplier: HONEYWELL SAFETY
Certificates
About this item
These fully reversible chainmail gloves are equipped with heavy duty adjustable straps and press stud fastening. They combines safety, quality and comfort, protecting against cuts, a common hazard in the meat and processing industries.
- Detachable and interchangeable plastic straps with antimicrobial agent
- Damaged straps can be changed over quickly and easily
- Firm fastening for a secure fit every time; there is no side slit which may allow a knife to enter at wrist level
- Fitted with an identification tag which can be personalised by the wearer
- Each glove is identified by a unique number for traceability and record keeping
- Exceptional quality of chain mesh welds, reducing the number of repairs, and increasing the life time of the glove
- Protection for hand and part of forearm
Typical applications include: Metal works, meat processing lines, kill floors, meat deboning, poultry works, fish processing.
Length: 393 - 482 mm (depending on size)
Weight: 309 - 412 g (depending on size)
Chainmail thickness: 0,05 mm
External diameter of ring: 4 mm
Caution: The colour given in the ordering table states the colour of the colour-coded plastic straps.
Specifications
- Catalog No:
- DALL2542315R0302
- DALL2542314R0302
- Ambidestro/mano specifico:
- ambidestro
- ambidestro
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- Tipo polsino:
- Lungo
- Lungo
- Approvati per il contatto con gli alimenti:
- +
- +