General purpose gloves, FineDex® 944-01 Clean
Supplier: HONEYWELL SAFETY
Certificates
About this item
These dipped latex gloves with cotton flock interior provide good mechanical and chemical resistance against detergents and liquid products.
- Liquidproof
- Natural latex provides excellent flexibility, dexterity and comfort
- Cotton flocked interior provides the user with greater comfort
- Diamond finish enables grasping of dry and damp objects
- Cuff helps to protect a large part of the forearm
- Production process helps to obtain a glove with a minimum latex protein
Ideal for the general maintenance of factories and collective areas, standard work in the chemical industry and collective catering.
Certifications: EN 388:2016 (1010X), EN ISO 374-1:2016 Type C (C), EN ISO 374-5:2016 (MICRO-ORGANISMS); Cat. II
Specifications
- Catalog No:
- DALL2094401/6
- DALL2094401/10
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- Materiale rivestimento interno:
- Cotone
- Cotone
- Finitura:
- Diamond-pattern
- Diamond-pattern
- Tipo polsino:
- Dentellato
- Dentellato
- Norma EN:
- EN 388:2016 (1010X), EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS)
- EN 388:2016 (1010X), EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS)