Overgloves, Electrosoft LT 2,5/5 kV
Nome del fornitore: HONEYWELL SAFETY
Certificati
Informazioni su questo articolo
These overgloves made from yellow cowhide grain leather are designed for the mechanical protection of low voltage 2,5 kV electrician's gloves (Electrosoft glove DALL2091903*) and 5 KV (Electrosoft CL 0 gloves).
- Silicone treatment ensures good flexibility, water repellency and good mechanical protection
- Gunn cut, thumb in one piece, gauntless
- Velcro adjustment strap on the back
- 150 mm split leather cuff sewed outside
- Protects Electrosoft gloves against risks of cuts and tearing
Certifications: EN 388:2016 (2121X); Cat. II
Specifiche
- Catalog No:
- DALL2012897/10
- DALL2012897/9
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- Tipo polsino:
- Svasato
- Svasato
- Lunghezza polsino:
- 150 mm
- 150 mm
- Norma EN:
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)