Pipette stand for Gilson pipettes, rotary, CARROUSEL™
Catalog # 612-1816
Supplier: GILSON
Specifications
- Colore:bianco
- Descrizione:Rotating pipette stand, CARROUSEL™, for 7 Gilson pipettes (except Pipetman Concept®)
- Conf:1
Specifications
About this item
This carousel stand holds up to seven single or multi channel Gilson pipettes.
- Robust and stable due to its heavy base
- Made of PP and PC: Strong resistance to aggressive chemicals
- Protects valuable instruments from shocks and contamination
- Pipettes hang vertically to prevent liquids from running inside the shaft
- Rotating head gives users an easy access to the pipette of their choice
- Space saving for a a neat, tidy and organised work bench