Pipette stand, rotary
Codice catalogo 613-5492
Nome del fornitore: Sartorius
Specifiche
- Per:6 mechanical pipettes
- Colore:bianco
- Descrizione:Carousel stand for 6 mechanical pipettes
- L×P×A:290×250×60 mm
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:Renewable energy certificate; ISO 14001
- Conf:1
Specifiche
Informazioni su questo articolo
Carousel stand for six mechanical pipettes.
When the pipette is not in use, it should be stored in an upright position in order to avoid contamination from desks and benches. This compact carousel stand is ideal for saving bench space and is compatible with all mechanical pipettes.