Weighing papers
76,2×76,2 mm
Codice catalogo HEATHS120116
Nome del fornitore: HEATHROW
Vai a
Product Details & Documents
These weighing papers are nitrogen-free for a very smooth surface and good sample recovery.
- Disposable
- Strong enough to support dense material
Colour: White
Specifiche
- Seq:01
- Taglia:76,2×76,2 mm