VWR®, Cap Mat Applicator
732-3335
- Descrizione:Cap mat applicator
- Seq:05
- Conf:1
Compact and portable, the cap mat applicator is very easy to use, requiring only one operation of the system to produce an accurate and tight seal on a wide range of both collection and shallow well microplates.
- Needs minimal pressure for capping, reducing fatigue and RSI
- Powder coated to resist chemical spillage