Wall-mounted cabinets
Made of epoxy powder coated steel, with one shelf.
- Solid hinged doors or sliding glass doors
- Height adjustable shelfs
- Maximum loading capacity: 80 kg
Made of epoxy powder coated steel, with one shelf.
- Solid hinged doors or sliding glass doors
- Height adjustable shelfs
- Maximum loading capacity: 80 kg
Product Family Options
- DescrizionePorteL×P×A
- Wall-mounted cabinet, with one shelf, lockable2 sliding glass doors900×350×480 mm1 * 1 STSFr. 960.00
- Wall- mounted cabinet, with one shelf, open front-900×350×480 mm1 * 1 STSFr. 340.00
- Wall-mounted cabinet, with one shelf, lockable2 sliding glass doors900×350×630 mm1 * 1 STSFr. 997.00
- Wall- mounted cabinet, with one shelf, open front-900×350×630 mm1 * 1 STSFr. 366.00
- Wall-mounted cabinet, with one shelf, lockable2 sliding glass doors900×350×780 mm1 * 1 STSFr. 1’040.00
- Wall- mounted cabinet, with one shelf, open front-900×350×780 mm1 * 1 STSFr. 391.00
- Wall-mounted cabinet, with one shelf2 doors900×366×480 mm1 * 1 STSFr. 612.00
- Wall-mounted cabinet, with one shelf, lockable2 doors900×366×480 mm1 * 1 STSFr. 676.00
- Wall-mounted cabinet, with one shelf2 doors900×366×630 mm1 * 1 STSFr. 666.00
- Wall-mounted cabinet, with one shelf, lockable2 doors900×366×630 mm1 * 1 STSFr. 732.00
- Wall-mounted cabinet, with one shelf2 doors900×366×780 mm1 * 1 STSFr. 714.00
- Wall-mounted cabinet, with one shelf, lockable2 doors900×366×780 mm1 * 1 STSFr. 781.00
- Wall-mounted cabinet, with one shelf, lockable2 sliding glass doors1200×350×480 mm1 * 1 STSFr. 1’070.00
- Wall- mounted cabinet, with one shelf, open front-1200×350×480 mm1 * 1 STSFr. 402.00
- Wall-mounted cabinet, with one shelf, lockable2 sliding glass doors1200×350×630 mm1 * 1 STSFr. 1’100.00
- Wall- mounted cabinet, with one shelf, open front-1200×350×630 mm1 * 1 STSFr. 422.00
- Wall-mounted cabinet, with one shelf, lockable2 sliding glass doors1200×350×780 mm1 * 1 STSFr. 1’180.00
- Wall- mounted cabinet, with one shelf, open front-1200×350×780 mm1 * 1 STSFr. 451.00
- Wall-mounted cabinet, with one shelf2 doors1200×366×480 mm1 * 1 STSFr. 691.00
- Wall-mounted cabinet, with one shelf, lockable2 doors1200×366×480 mm1 * 1 STSFr. 758.00
- Wall-mounted cabinet, with one shelf, lockable2 doors1200×366×630 mm1 * 1 STSFr. 804.00
- Wall-mounted cabinet, with one shelf2 doors1200×366×630 mm1 * 1 STSFr. 738.00
- Wall-mounted cabinet, with one shelf2 doors1200×366×780 mm1 * 1 STSFr. 808.00
- Wall-mounted cabinet, with one shelf, lockable2 doors1200×366×780 mm1 * 1 STSFr. 875.00
- Wall-mounted cabinet, with one shelf3 doors1500×350×480 mm1 * 1 STSFr. 1’090.00
- Wall-mounted cabinet, with one shelf, lockable2 sliding glass doors1500×350×480 mm1 * 1 STSFr. 1’190.00
- Wall- mounted cabinet, with one shelf, open front-1500×350×480 mm1 * 1 STSFr. 461.00
- Wall-mounted cabinet, with one shelf, lockable3 doors1500×350×480 mm1 * 1 STSFr. 1’170.00
- Wall-mounted cabinet, with one shelf, lockable2 sliding glass doors1500×350×630 mm1 * 1 STSFr. 1’270.00
- Wall- mounted cabinet, with one shelf, open front-1500×350×630 mm1 * 1 STSFr. 493.00
- Wall-mounted cabinet, with one shelf, lockable3 doors1500×350×630 mm1 * 1 STSFr. 1’120.00
- Wall-mounted cabinet, with one shelf3 doors1500×350×630 mm1 * 1 STSFr. 1’040.00
- Wall-mounted cabinet, with one shelf, lockable2 sliding glass doors1500×350×780 mm1 * 1 STSFr. 1’360.00
- Wall- mounted cabinet, with one shelf, open front-1500×350×780 mm1 * 1 STSFr. 536.00
- Wall-mounted cabinet, with one shelf3 doors1500×350×780 mm1 * 1 STSFr. 1’130.00
- Wall-mounted cabinet, with one shelf, lockable3 doors1500×350×780 mm1 * 1 STSFr. 1’210.00