Apron with sleeves, MICROCHEM® 3000, model 215
ANSEYE30W-00215-02
: Ansell
- Taglia dell'abbigliamento:S
- Applicazioni per abiti:Chemical protection
- Colore:giallo
- Seq:01
- Conf:30
This apron is made from an external film barrier layer with two non woven layers underneath. The fabric provides a strong liquid and particle barrier.
- Velcro fastening to neck Tie fastening at the waist Double cuff with knitted inner cuff Welded and taped seams Multi-layer barrier fabric is effective against numerous chemicals Highly visible: Bright yellow for improved worker safety Lightweight yet durable
Typical applications include: Chemicals, oil and petrochemicals, pharmaceutical, food industry (caustic clean downs), sewage purification installations, industrial and tank cleaning, mining
: Type 3-B/4-B/5-B, EN 14126, EN 1073-2, EN 1149-5