Micro injection port adapter assembly, Upchurch Scientific®
Catalog # UPCHM-432-03
Supplier: UPCHURCH SCIENTIFIC
Specifications
- Descrizione:Replacement tubing/fitting assembly
- Seq:02
- Conf:1
Specifications
About this item
This micro injection port adapter assembly can be connected to 360 µm Ext.Ø capillary tubing to introduce samples. It accepts standard 22 gauge Hamilton-style injection syringe needles. The needle butts directly against the connecting tubing during injections, therefore no additional swept volume is added to the fluid pathway by this adapter.
- For 360 µm Ext.Ø. tubing
- Mount on bracket or bulkhead