Syrup bottles, narrow neck
Catalog # ALUG1000019
Supplier: APG Europe
Specifications
- Ø est:46,1 mm
- Tipo:Alpha
- Capacità:100 ml
- Altezza:105 mm
- Conf:86
Specifications
About this item
These ready to fill syrup bottles are made of type 3 clear glass with screw neck PP 28.
- Pharmaceutical grade moulded glass
- SGD mold reference 52473
- Light transmission for amber glass is full compliance with USP and EP
- Clean, particulate and microbiological control
- Type: alpha