Bottles, narrow neck, alpha, lightweight with screw caps
Catalog # 215-2119
Anbieter: Azpack
Technische Daten
- Ø est:46,00 mm
- Filettatura:28 mm
- Capacità:100 ml
- Colore:trasparente
- Altezza:105,00 mm
- Seq:21
- Conf:20
Technische Daten
Über diesen Artikel
Clear or amber lightweight glass.
- For use in applications without high thermal loads
- Amber coloured to protect light-sensitive material
- A lighter, stronger and more uniform range of bottles
Large range of standard round glass bottles. All sizes of the lightweight bottles have exactly the same neck which means that only one cap size for the entire range has to be stocked.