Safety spectacles, Solis
Anti-UV (5-3,2), anti-scratch, polarised coating, nero, Smoke PC
Codice catalogo 111-0156
Nome del fornitore: Bollé Safety
Vai a
Product Details & Documents
Solis spectacles offer a wrap-around style with the benefits of development of modern materials.
- Non slip TIPGRIP temples
- Non slip bridge
- Integral side shields
- Polarised coating
Weight: 25 g
Certifications: EN 166, EN 172
Informazioni per l'ordine: Supplied with microfibre pouch.
Specifiche
- Tipo:Anti-UV (5-3,2), anti-scratch, polarised coating
- Colore:nero
- Materiale lente:Smoke PC