Cleanroom boot covers, BioClean-D™, CleanTough™
Supplier: Nitritex
Certificates
About this item
These disposable boot covers are made of BioClean-D™, a lightweight, low linting and hard-wearing fabric (PP laminated with a PE film) which provides maximum comfort and protects against fine sprays and particles.
- Slip resistant Safe-Step™ sole enhances wearer safety
- Available as sterile or non sterile version
- Elasticated top with secure ties
- A unique barcode on the outer carton (ITF-14) allows automatic data entry into Stock Control Systems
Suitable for use in Class 10, ISO 4, M2.5 environments.
Length: 400 mm
Certifications: EN ISO 13034 (Type PB[6]), EN 14325, EN 1149-5, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2; Cat. III
Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Sterile model: Sterilised in accordance with ISO 11137-1:2006.
Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Sterile model: Sterilised in accordance with ISO 11137-1:2006.
Packaging: Sterile version: One pair per sealed plastic bag, 15 sealed bags per sealed outer bag, five sealed bags of 15 pairs per sealed carton liner, one carton liner per carton (75 pairs).
Non sterile version: 5 bags of 30 pieces per sealed carton liner; one carton liner per carton (150 pieces).
Non sterile version: 5 bags of 30 pieces per sealed carton liner; one carton liner per carton (150 pieces).
Specifications
- Catalog No:
- 113-2265
- 113-0814
- Classe di cleanroom:
- 10/ISO 4
- 10/ISO 4
- Applicazioni per abiti:
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
Specifications
Related Information
Frequently Bought Together