ESD shoe covers, BioClean ESD™
: Nitritex
The anti-static ESD disposable overshoes with conductive tape are manufactured from non woven spunbonded polypropylene.
- Superior ESD performance
- Fitted with a polyester conductive ribbon with a conductive carbon filament
- Non marking ribbon
- Low levels of particulate shedding and double bagged for cleanroom use
Suitable for use in ISO class 5 cleanrooms.
Length: 400 or 455 mm
- Catalog No:
- 113-0815
- 113-0816
- 113-0817
- Classe di cleanroom:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Applicazioni per abiti:
- Cleanroom
- Cleanroom
- Cleanroom
Frequently Bought Together