Orders LinkContactUsLinkComponent
High visibility jacket, Strada 2 High-Visibility
Nome del fornitore:   DELTA PLUS
High visibility jacket, Strada 2 High-Visibility
Nome del fornitore:  DELTA PLUS
Certificates
High visibility jacket, Strada 2 High-Visibility
2 varianti
Product Details & Documents
This high-visibility safety jacket (class 3) is made from high visibility, fluorescent polyester with internal polyurethane coating and polyester lining. The jacket includes a zip with press stud cover, two pockets with press stud pocket covers, two side pockets, an inside pocket, and waterproof seams. Suitable for ∼50 washes at 40 °C.
  • Two reflective strips on each sleeve and strips on the body and shoulders
  • Non detachable hood with velcro fastening, which can be folded in the collar
  • Straight sleeve openings with wind-resistant, internal elastic cuffs
  • Ventilation eyelets in the underarm area
  • Can be tightened at the waist with elastic cord and toggle
Colour: Fluorescent yellow/marine blue or fluorescent orange/marine blue.
 : EN 340; EN 342; EN 343; EN 471
Specifications for Product Family
Specifiche
  • Catalog No:
  • 113-0914
  • 113-0913
  • Applicazioni per abiti:
  • Industrial
  • Industrial
  • Risk area:
  • HiVis
  • HiVis
  • Genere:
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Tipo di chiusura:
  • Con chiusura a zip
  • Con chiusura a zip
  • Tessuto:
  • Polyester with internal polyurethane coating
  • Polyester with internal polyurethane coating
Specifiche
Product Family Options
Informazioni sul prodotto
  • Colore
    Taglia dell'abbigliamento
    Disponibilità
    Price
  • arancione/blu
    L
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 154.00
    SFr. 154.00
    1 * 1 ST
       SFr. 154.00
    Specifiche:
    More
    Specifications
    Cat. No.
    113-0914
    Applicazioni per abiti
    Industrial
    Risk area
    HiVis
    Genere
    Unisex
    Sleeve or leg length
    Long
    Tipo di chiusura
    Con chiusura a zip
    Tessuto
    Polyester with internal polyurethane coating
  • giallo/blu
    L
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 154.00
    SFr. 154.00
    1 * 1 ST
       SFr. 154.00
    Specifiche:
    More
    Specifications
    Cat. No.
    113-0913
    Applicazioni per abiti
    Industrial
    Risk area
    HiVis
    Genere
    Unisex
    Sleeve or leg length
    Long
    Tipo di chiusura
    Con chiusura a zip
    Tessuto
    Polyester with internal polyurethane coating