Adhesive seal, VIEWseal™
VIEWseal™, Transparent
Codice catalogo 736-0232
Nome del fornitore: Greiner Bio-One
Adhesive seal, VIEWseal™
Codice catalogo 736-0232
Nome del fornitore: Greiner Bio-One
Vai a
Product Details & Documents
VIEWseal™ is an adhesive sealing film coated with a silicone adhesive, which only sticks when the film surface is pressed on. The film is easy to work with, even when wearing gloves (the film does not adhere to gloves).
- Temperature resistant from −80 to 110 °C
- Highly transparent for precise optical measurements
- Ideally suited for protein crystallography
Specifiche
- Proprietà ottica:Transparent
- Descrizione:VIEWseal™
- Cat. No.:736-0232
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:ISO 14001 (241002); ISO 50001 (241002)