Dehydrated culture media, Terrific broth II, MP Biomedicals
Codice catalogo ICNA113046075
Nome del fornitore: MP Biomedicals
Numero CAS:
Specifiche
- Conf:10 L
- Descrizione:Terrific broth II, pack pouches
- Seq:07
- Volume:10000 ml
Specifiche
Informazioni su questo articolo
Terrific broth (TB) has been formulated for optimum growth of Escherichia coli for purposes of strain maintenance, cloning, protein expression. It is certified for plasmid and other bacterial cultures in liquid medium. Higher cell densities are achieved using this broth.
- Molecular biology certified bacterial medium
Terrific Broth II is a proprietary formulation that is shipped as a single powder instead of as two separate components exhibiting same growth characteristics as the original terrific broth formula.